Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human Frizzled-7 FZD7 Protein, Fc Tag,Human Frizzled-7, Fc Tag (FZ7-H5258) is expressed from human 293 cells (HEK 293). It contains AA Gln 33 - Leu 185 (Accession AAH15915).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 100 µG
Catalog # ACRBFZ7-H5258100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Frizzled-7 FZD7 Protein, Fc Tag,Human Frizzled-7, Fc Tag (FZ7-H5258) is expressed from human 293 cells (HEK 293). It contains AA Gln 33 - Leu 185 (Accession AAH15915).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 100 µG
Catalog # ACRBFZ7-H5258100UG
Supplier:  ACRO Biosystems
Frequently Bought Together