Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Rhesus macaque Siglec-2 CD22 Protein, His Tag,Rhesus macaque Siglec-2, His Tag (SI2-R52Ha) is expressed from huma n 293 cells (HEK293). It contains AA Ser 20 - Arg 686 (Accession EHH29920.1).This protein carries a polyhistidine tag 1 * 1 mg
Catalog # ACRBSI2-R52HA-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Rhesus macaque Siglec-2 CD22 Protein, His Tag,Rhesus macaque Siglec-2, His Tag (SI2-R52Ha) is expressed from huma n 293 cells (HEK293). It contains AA Ser 20 - Arg 686 (Accession EHH29920.1).This protein carries a polyhistidine tag 1 * 1 mg
Catalog # ACRBSI2-R52HA-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together