Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human Fetuin B FETUA Protein, His Tag,Human Fetuin B, His Tag (FEB-H52H7) is expressed from human 293 cells (HEK2 93). It contains AA Cys 16 - Pro 382 (Accession AAH69670 ).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg
Catalog # ACRBFEB-H52H7-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Fetuin B FETUA Protein, His Tag,Human Fetuin B, His Tag (FEB-H52H7) is expressed from human 293 cells (HEK2 93). It contains AA Cys 16 - Pro 382 (Accession AAH69670 ).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg
Catalog # ACRBFEB-H52H7-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together