Order Entry
Netherlands
ContactUsLinkComponent
Mouse ITGAVITGB8 Heterodimer Protein, His TagTag Free (IT8-M52W6) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 988 (ITGAV) Gly 22 - Leu 681 (ITGB8) (Accession P43406-1 (ITGAV) Q0VBD0-1 (ITGB8)). 1 * 100 µG
Catalog # ACRBIT8-M52W6100UG
Supplier:  ACRO Biosystems
Mouse ITGAVITGB8 Heterodimer Protein, His TagTag Free (IT8-M52W6) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 988 (ITGAV) Gly 22 - Leu 681 (ITGB8) (Accession P43406-1 (ITGAV) Q0VBD0-1 (ITGB8)). 1 * 100 µG
Catalog # ACRBIT8-M52W6100UG
Supplier:  ACRO Biosystems
Frequently Bought Together