Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human PSMA FOLH1 Protein, His Tag,Human PSMA, His Tag (PSA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession Q04609-1 ).This protein carries a polyhistidine tag at the N-terminus. The pro 1 * 1 mg
Catalog # ACRBPSA-H52H3-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human PSMA FOLH1 Protein, His Tag,Human PSMA, His Tag (PSA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession Q04609-1 ).This protein carries a polyhistidine tag at the N-terminus. The pro 1 * 1 mg
Catalog # ACRBPSA-H52H3-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together