Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Mouse CD3 epsilon Protein, Fc Tag,Mouse CD3E, Fc Tag (CDE-M5256) is expressed from human 293 cells (HEK293). It c ontains AA Asp 23 - Asp 108 (Accession P22646).This protein carries a human IgG1 Fc tag at the C-terminus. The protei 1 * 100 µG
Catalog # ACRBCDE-M5256100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Mouse CD3 epsilon Protein, Fc Tag,Mouse CD3E, Fc Tag (CDE-M5256) is expressed from human 293 cells (HEK293). It c ontains AA Asp 23 - Asp 108 (Accession P22646).This protein carries a human IgG1 Fc tag at the C-terminus. The protei 1 * 100 µG
Catalog # ACRBCDE-M5256100UG
Supplier:  ACRO Biosystems
Frequently Bought Together