Order Entry
Netherlands
ContactUsLinkComponent
Human LAG-3, Llama IgG2b Fc Tag, low endotoxin (LA3-H525c) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 4 50 (Accession P18627-1). 1 * 1 mg
Catalog # ACRBLA3-H525C-1MG
Supplier:  ACRO Biosystems
Human LAG-3, Llama IgG2b Fc Tag, low endotoxin (LA3-H525c) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 4 50 (Accession P18627-1). 1 * 1 mg
Catalog # ACRBLA3-H525C-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together