Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human SECTM1 Protein, His Tag,Human SECTM1, His Tag (SE1-H5227) is expressed from human 293 cells (HEK293). It co ntains AA Gln 29 - Gly 145 (Accession AAH17716).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 1 mg
Catalog # ACRBSE1-H5227-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human SECTM1 Protein, His Tag,Human SECTM1, His Tag (SE1-H5227) is expressed from human 293 cells (HEK293). It co ntains AA Gln 29 - Gly 145 (Accession AAH17716).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 1 mg
Catalog # ACRBSE1-H5227-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together