Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Rabbit M-CSF R CSF1R CD115 Protein, His Tag,Rabbit M-CSF R, His Tag (CSR-R82H5) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Ser 503 (Accession G1TTE0-1).This protein carries a polyhistidine tag at the C-ter 1 * 100 µG
Catalog # ACRBCSR-R82H5100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Rabbit M-CSF R CSF1R CD115 Protein, His Tag,Rabbit M-CSF R, His Tag (CSR-R82H5) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Ser 503 (Accession G1TTE0-1).This protein carries a polyhistidine tag at the C-ter 1 * 100 µG
Catalog # ACRBCSR-R82H5100UG
Supplier:  ACRO Biosystems
Frequently Bought Together