Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Mouse CD38 Protein, His Tag,Mouse CD38, His Tag (CD8-M5223) is expressed from human 293 cells (HEK293). It contai ns AA Leu 45 - Thr 304 (Accession P56528-1).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 100 µG
Catalog # ACRBCD8-M5223100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Mouse CD38 Protein, His Tag,Mouse CD38, His Tag (CD8-M5223) is expressed from human 293 cells (HEK293). It contai ns AA Leu 45 - Thr 304 (Accession P56528-1).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 100 µG
Catalog # ACRBCD8-M5223100UG
Supplier:  ACRO Biosystems
Frequently Bought Together