Recombinat Protein,Mouse gp130 CD130 IL-6 R beta Protein, Fc Tag,Mouse gp130, Fc Tag (ILT-M5252) is expressed from human 293 cells ( HEK293). It contains AA Gln 23 - Glu 617 (Accession NP034690).This protein carries a human IgG1 Fc tag at the C-termi 1 * 100 µG
Catalog # ACRBILT-M5252100UG
Supplier: ACRO Biosystems
Frequently Bought Together