Bestelling invoeren
Netherlands
ContactUsLinkComponent
Mouse ICOS (C137S, C138S), His Tag (ICS-M52H8) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Leu 144 (Accessio n Q9WVS0-1 (C137S,C138S)). 1 * 1 mg
Catalog # ACRBICS-M52H8-1MG
Supplier:  ACRO Biosystems
Mouse ICOS (C137S, C138S), His Tag (ICS-M52H8) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Leu 144 (Accessio n Q9WVS0-1 (C137S,C138S)). 1 * 1 mg
Catalog # ACRBICS-M52H8-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together