Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human PTH1R PTHR1 Protein, His Tag,Human PTH1R, His Tag (PTR-H5222) is expressed from human 293 cells (HEK293). I t contains AA Asp 27 - Gly 188 (Accession NP000307).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 100 µG
  ACRBPTR-H5222100UG
Recombinat Protein,Human PTH1R PTHR1 Protein, His Tag,Human PTH1R, His Tag (PTR-H5222) is expressed from human 293 cells (HEK293). I t contains AA Asp 27 - Gly 188 (Accession NP000307).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 100 µG
  ACRBPTR-H5222100UG
Frequently Bought Together