Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human TWEAK R TNFRSF12A Protein, Fc Tag,Human TWEAK R, Fc Tag (TNA-H526a) is expressed from human 293 cells (HEK2 93). It contains AA Glu 28 - Trp 79 (Accession AAH02718).This protein carries a human IgG1 Fc tag at the N-terminus. 1 * 100 µG
Catalog # ACRBTNA-H526A100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human TWEAK R TNFRSF12A Protein, Fc Tag,Human TWEAK R, Fc Tag (TNA-H526a) is expressed from human 293 cells (HEK2 93). It contains AA Glu 28 - Trp 79 (Accession AAH02718).This protein carries a human IgG1 Fc tag at the N-terminus. 1 * 100 µG
Catalog # ACRBTNA-H526A100UG
Supplier:  ACRO Biosystems
Frequently Bought Together