Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Rhesus macaque CD4 Protein, His Tag,Rhesus macaque CD4, His Tag (CD4-C52H7) is expressed from human 293 cells (HE K293). It contains AA Lys 26 - Trp 390 (Accession G7N5T8).This protein carries a polyhistidine tag at the C-terminus. 1 * 100 µG
Catalog # ACRBCD4-C52H7100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Rhesus macaque CD4 Protein, His Tag,Rhesus macaque CD4, His Tag (CD4-C52H7) is expressed from human 293 cells (HE K293). It contains AA Lys 26 - Trp 390 (Accession G7N5T8).This protein carries a polyhistidine tag at the C-terminus. 1 * 100 µG
Catalog # ACRBCD4-C52H7100UG
Supplier:  ACRO Biosystems
Frequently Bought Together