Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human VSIG2 Protein, His Tag,Human VSIG2, His Tag (VS2-H5225) is expressed from human 293 cells (HEK293). It cont ains AA Val 24 - Ala 243 (Accession AAH07313).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 1 mg
Catalog # ACRBVS2-H5225-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human VSIG2 Protein, His Tag,Human VSIG2, His Tag (VS2-H5225) is expressed from human 293 cells (HEK293). It cont ains AA Val 24 - Ala 243 (Accession AAH07313).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 1 mg
Catalog # ACRBVS2-H5225-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together