Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Biotinylated Cynomolgus Her2 ErbB2 Protein, His,Avitag™,Biotinylated Cynomolgus Her2, His,Avitag (HE2-C82E3) is e xpressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession XP005584091.2).This protein carrie 1 * 200 µG
Catalog # ACRBHE2-C82E3200UG
Supplier:  ACRO Biosystems
Recombinat Protein,Biotinylated Cynomolgus Her2 ErbB2 Protein, His,Avitag™,Biotinylated Cynomolgus Her2, His,Avitag (HE2-C82E3) is e xpressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession XP005584091.2).This protein carrie 1 * 200 µG
Catalog # ACRBHE2-C82E3200UG
Supplier:  ACRO Biosystems
Frequently Bought Together