Recombinat Protein,Human Siglec-6 CD327 Protein, Fc Tag,Human Siglec-6, Fc Tag (SI6-H5256) is expressed from human 293 cells (HEK293 ). It contains AA Gln 16 - Val 320 (Accession AAH35359).This protein carries a human IgG1 Fc tag at the C-terminus. T 1 * 100 µG
Catalog # ACRBSI6-H5256100UG
Leverancier: ACRO Biosystems
Frequently Bought Together