Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Mouse HMGB1 Protein, His Tag,Mouse HMGB1, His Tag (HM1-M52H6) is expressed from human 293 cells (HEK293). It cont ains AA Met 1 - Glu 215 (Accession P63158-1).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 100 µG
Catalog # ACRBHM1-M52H6100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Mouse HMGB1 Protein, His Tag,Mouse HMGB1, His Tag (HM1-M52H6) is expressed from human 293 cells (HEK293). It cont ains AA Met 1 - Glu 215 (Accession P63158-1).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 100 µG
Catalog # ACRBHM1-M52H6100UG
Supplier:  ACRO Biosystems
Frequently Bought Together