Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Mouse CD47 Protein, Fc Tag,Mouse CD47, Fc Tag (CD7-M5251) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession Q61735).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a 1 * 1 mg
Catalog # ACRBCD7-M5251-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Mouse CD47 Protein, Fc Tag,Mouse CD47, Fc Tag (CD7-M5251) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession Q61735).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a 1 * 1 mg
Catalog # ACRBCD7-M5251-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together