Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Mouse LAG-3 CD223 Protein, His Tag,Mouse LAG-3, His Tag (LA3-M52H5) is expressed from human 293 cells (HEK293). I t contains AA Gly 24 - Leu 442 (Accession Q61790).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 100 µG
Catalog # ACRBLA3-M52H5100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Mouse LAG-3 CD223 Protein, His Tag,Mouse LAG-3, His Tag (LA3-M52H5) is expressed from human 293 cells (HEK293). I t contains AA Gly 24 - Leu 442 (Accession Q61790).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 100 µG
Catalog # ACRBLA3-M52H5100UG
Supplier:  ACRO Biosystems
Frequently Bought Together