Order Entry
Netherlands
ContactUsLinkComponent
Human ITGAVITGB1 Heterodimer Protein, His TagTag Free (IT1-H52E1) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) Gln 21 - Asp 728 (ITGB1) (Accession NP002201.1 (ITGAV) NP002202.2 (ITGB1)). 1 * 100 µG
Catalog # ACRBIT1-H52E1100UG
Supplier:  ACRO Biosystems
Human ITGAVITGB1 Heterodimer Protein, His TagTag Free (IT1-H52E1) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) Gln 21 - Asp 728 (ITGB1) (Accession NP002201.1 (ITGAV) NP002202.2 (ITGB1)). 1 * 100 µG
Catalog # ACRBIT1-H52E1100UG
Supplier:  ACRO Biosystems
Frequently Bought Together