Recombinat Protein,Human CD94 Protein, Mouse IgG2a Fc Tag,Human CD94, Mouse IgG2a Fc Tag (CD4-H5253) is expressed from human 293 cel ls (HEK293). It contains AA Lys 32 - Ile 179 (Accession Q13241-1).This protein carries a mouse IgG2a Fc tag at the C- 1 * 100 µG
Catalog # ACRBCD4-H5253100UG
Supplier: ACRO Biosystems
Frequently Bought Together