Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Mouse IgG2a Fc Protein, Tag Free,Mouse IgG Fc, Tag Free (IGA-M5207) is expressed from human 293 cells (HEK293). I t contains AA Glu 98 - Lys 330 (Accession CAC20702).This protein carries no tag. The protein has a calculated MW of 2 1 * 200 µG
Catalog # ACRBIGA-M5207200UG
Supplier:  ACRO Biosystems
Recombinat Protein,Mouse IgG2a Fc Protein, Tag Free,Mouse IgG Fc, Tag Free (IGA-M5207) is expressed from human 293 cells (HEK293). I t contains AA Glu 98 - Lys 330 (Accession CAC20702).This protein carries no tag. The protein has a calculated MW of 2 1 * 200 µG
Catalog # ACRBIGA-M5207200UG
Supplier:  ACRO Biosystems
Frequently Bought Together