Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human DLL3 Protein, His Tag,Human DLL3, His Tag (DL3-H52H4) is expressed from human 293 cells (HEK293). It contai ns AA Ala 27 - Leu 492 (Accession Q9NYJ7-1).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 100 µG
Catalog # ACRBDL3-H52H4100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human DLL3 Protein, His Tag,Human DLL3, His Tag (DL3-H52H4) is expressed from human 293 cells (HEK293). It contai ns AA Ala 27 - Leu 492 (Accession Q9NYJ7-1).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 100 µG
Catalog # ACRBDL3-H52H4100UG
Supplier:  ACRO Biosystems
Frequently Bought Together