Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Mouse TIM-3 HAVCR2 Protein, His Tag,Mouse TIM-3, His Tag (TI3-M52H1) is expressed from human 293 cells (HEK293). It contains AA Arg 20 - Arg 191 (Accession AAL65156.1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 100 µG
Catalog # ACRBTI3-M52H1100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Mouse TIM-3 HAVCR2 Protein, His Tag,Mouse TIM-3, His Tag (TI3-M52H1) is expressed from human 293 cells (HEK293). It contains AA Arg 20 - Arg 191 (Accession AAL65156.1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 100 µG
Catalog # ACRBTI3-M52H1100UG
Supplier:  ACRO Biosystems
Frequently Bought Together