Recombinat Protein,Rabbit PD-1 PDCD1 Protein, His Tag,Rabbit PD-1, His Tag (PD1-R52H0) is expressed from human 293 cells (HEK293). I t contains AA Leu 25 - Gln 167 (Accession G1SUF0).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 100 µG
Catalog # ACRBPD1-R52H0100UG
Supplier: ACRO Biosystems
Frequently Bought Together