Order Entry
Netherlands
ContactUsLinkComponent
Biotinylated Human IL-17AIL-17F, Avitag,His Tag (ILF-H82W1) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (IL-17A) Arg 31 - Gln 163 (IL-17F) (Accession Q16552-1 (IL-17A) Q96PD4-1 (IL-17F)). 1 * 200 µG
Catalog # ACRBILF-H82W1200UG
Supplier:  ACRO Biosystems
Biotinylated Human IL-17AIL-17F, Avitag,His Tag (ILF-H82W1) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (IL-17A) Arg 31 - Gln 163 (IL-17F) (Accession Q16552-1 (IL-17A) Q96PD4-1 (IL-17F)). 1 * 200 µG
Catalog # ACRBILF-H82W1200UG
Supplier:  ACRO Biosystems
Frequently Bought Together