Order Entry
Netherlands
ContactUsLinkComponent
Human ITGA9ITGB1 Heterodimer Protein, His TagTag Free (IT1-H52W4) is expressed from human 293 cells (HEK293). It contains AA Tyr 30 - Trp 981 (ITGA9) Gln 21 - Asp 728 (ITGB1) (Accession Q13797-1 (ITGA9) P05556-1 (ITGB1)). 1 * 100 µG
Catalog # ACRBIT1-H52W4100UG
Supplier:  ACRO Biosystems
Human ITGA9ITGB1 Heterodimer Protein, His TagTag Free (IT1-H52W4) is expressed from human 293 cells (HEK293). It contains AA Tyr 30 - Trp 981 (ITGA9) Gln 21 - Asp 728 (ITGB1) (Accession Q13797-1 (ITGA9) P05556-1 (ITGB1)). 1 * 100 µG
Catalog # ACRBIT1-H52W4100UG
Supplier:  ACRO Biosystems
Frequently Bought Together