Recombinat Protein,Mouse LIF Protein, His Tag,Mouse LIF, His Tag (LIF-M5227) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Phe 203 (Accession P09056).This protein carries a polyhistidine tag at the C-terminus. The protein has a 1 * 50 µG
Catalog # ACRBLIF-M5227-50UG
Supplier: ACRO Biosystems
Frequently Bought Together