Recombinat Protein,Mouse IL-10 Protein, His Tag,Mouse IL-10, His Tag (IL0-M4248) is expressed from human 293 cells (HEK293). It cont ains AA Ser 19 - Ser 178 (Accession NP034678).This protein carries a polyhistidine tag at the N-terminus. The protein 1 * 20 µG
Catalog # ACRBIL0-M4248-20UG
Supplier: ACRO Biosystems
Frequently Bought Together