Order Entry
Netherlands
ContactUsLinkComponent
Human Siglec-3, Llama IgG2b Fc Tag, low endotoxin (CD3-H5259) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - Hi s 259 (Accession AAH28152.1). 1 * 100 µG
Catalog # ACRBCD3-H5259100UG
Supplier:  ACRO Biosystems
Human Siglec-3, Llama IgG2b Fc Tag, low endotoxin (CD3-H5259) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - Hi s 259 (Accession AAH28152.1). 1 * 100 µG
Catalog # ACRBCD3-H5259100UG
Supplier:  ACRO Biosystems
Frequently Bought Together