Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human IL-18BP Protein, His Tag,Human IL-18BP, His Tag (ILP-H5222) is expressed from human 293 cells (HEK293). It contains AA Thr 31 - Gly 194 (Accession NP001138529).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 200 µG
Catalog # ACRBILP-H5222200UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human IL-18BP Protein, His Tag,Human IL-18BP, His Tag (ILP-H5222) is expressed from human 293 cells (HEK293). It contains AA Thr 31 - Gly 194 (Accession NP001138529).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 200 µG
Catalog # ACRBILP-H5222200UG
Supplier:  ACRO Biosystems
Frequently Bought Together