Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human IL-18 R1 CD218a Protein, Fc Tag,Human IL-18 R1, Fc Tag (IL1-H5259) is expressed from human 293 cells (HEK29 3). It contains AA Ala 19 - Arg 329 (Accession AAH69575).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 1 mg
Catalog # ACRBIL1-H5259-1MG
Leverancier:  ACRO Biosystems
Recombinat Protein,Human IL-18 R1 CD218a Protein, Fc Tag,Human IL-18 R1, Fc Tag (IL1-H5259) is expressed from human 293 cells (HEK29 3). It contains AA Ala 19 - Arg 329 (Accession AAH69575).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 1 mg
Catalog # ACRBIL1-H5259-1MG
Leverancier:  ACRO Biosystems
Frequently Bought Together