Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human CLEC12A MICL CLL-1 Protein, Fc Tag,Human CLEC12A, Fc Tag (CLA-H5266) is expressed from human 293 cells (HEK 293). It contains AA His 65 - Ala 265 (Accession Q5QGZ9-2).This protein carries a human IgG1 Fc tag at the N-terminus 1 * 50 µG
  ACRBCLA-H5266-50UG
Recombinat Protein,Human CLEC12A MICL CLL-1 Protein, Fc Tag,Human CLEC12A, Fc Tag (CLA-H5266) is expressed from human 293 cells (HEK 293). It contains AA His 65 - Ala 265 (Accession Q5QGZ9-2).This protein carries a human IgG1 Fc tag at the N-terminus 1 * 50 µG
  ACRBCLA-H5266-50UG
Frequently Bought Together