Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Mouse PSMA FOLH1 Protein, His Tag,Mouse PSMA, His Tag (PSA-M5245) is expressed from human 293 cells (HEK293). It contains AA Ile 44 - Ala 752 (Accession O35409-1).This protein carries a polyhistidine tag at the N-terminus. The prot 1 * 1 mg
Catalog # ACRBPSA-M5245-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Mouse PSMA FOLH1 Protein, His Tag,Mouse PSMA, His Tag (PSA-M5245) is expressed from human 293 cells (HEK293). It contains AA Ile 44 - Ala 752 (Accession O35409-1).This protein carries a polyhistidine tag at the N-terminus. The prot 1 * 1 mg
Catalog # ACRBPSA-M5245-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together