Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human GDF-15 MIC-1 Protein, His Tag,Human GDF-15, His Tag (GD5-H5149) is expressed from E.coli cells. It contains AA Ala 197 - Ile 308 (Accession Q99988-1).This protein carries a polyhistidine tag at the N-terminus. The protein ha 1 * 100 µG
Catalog # ACRBGD5-H5149100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human GDF-15 MIC-1 Protein, His Tag,Human GDF-15, His Tag (GD5-H5149) is expressed from E.coli cells. It contains AA Ala 197 - Ile 308 (Accession Q99988-1).This protein carries a polyhistidine tag at the N-terminus. The protein ha 1 * 100 µG
Catalog # ACRBGD5-H5149100UG
Supplier:  ACRO Biosystems
Frequently Bought Together