Recombinat Protein,Cynomolgus TSLP (R127A, R130S) Protein, His Tag,Cynomolgus TSLP (R127A, R130S), His Tag (TSP-C52H4) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession A0A2K5TXV0-1 (R127A, R130S)).This protein ca 1 * 1 mg
Catalog # ACRBTSP-C52H4-1MG
Supplier: ACRO Biosystems
Frequently Bought Together