Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human Glypican 2 GPC2 Protein, Fc Tag,Human Glypican 2, Fc Tag (GP2-H5255) is expressed from human 293 cells (HEK 293). It contains AA Ser 24 - Ser 553 (Accession Q8N158-1).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 100 µG
Catalog # ACRBGP2-H5255100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Glypican 2 GPC2 Protein, Fc Tag,Human Glypican 2, Fc Tag (GP2-H5255) is expressed from human 293 cells (HEK 293). It contains AA Ser 24 - Ser 553 (Accession Q8N158-1).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 100 µG
Catalog # ACRBGP2-H5255100UG
Supplier:  ACRO Biosystems
Frequently Bought Together