Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human Neprilysin MME CD10 Protein, Fc Tag,Human Neprilysin, Fc Tag (MME-H526a) is expressed from human 293 cells (HEK293). It contains AA Tyr 52 - Trp 750 (Accession NP000893.2).This protein carries a human IgG1 Fc tag at the N-ter 1 * 50 µG
Catalog # ACRBMME-H526A-50UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Neprilysin MME CD10 Protein, Fc Tag,Human Neprilysin, Fc Tag (MME-H526a) is expressed from human 293 cells (HEK293). It contains AA Tyr 52 - Trp 750 (Accession NP000893.2).This protein carries a human IgG1 Fc tag at the N-ter 1 * 50 µG
Catalog # ACRBMME-H526A-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together