Order Entry
Netherlands
ContactUsLinkComponent
HIV-1 HIV-1Clade BC (CN54) GP120, His Tag derived from the env. gene of HIV-1 strain CN54 gp160 (Accession G4XFJ5-1 (E46G, T396A, A4 97T), Thr 36 - Arg 507) and glycosylated with N-linked sugars and expressed in HEK293 cells at ACRObiosystems. 1 * 100 µG
Catalog # ACRBGP4V15227100UG
Supplier:  ACRO Biosystems
HIV-1 HIV-1Clade BC (CN54) GP120, His Tag derived from the env. gene of HIV-1 strain CN54 gp160 (Accession G4XFJ5-1 (E46G, T396A, A4 97T), Thr 36 - Arg 507) and glycosylated with N-linked sugars and expressed in HEK293 cells at ACRObiosystems. 1 * 100 µG
Catalog # ACRBGP4V15227100UG
Supplier:  ACRO Biosystems
Frequently Bought Together