Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human BTLA (31-150) Protein, Mouse IgG2a Fc Tag,Human BTLA (31-150), Mouse IgG2a Fc Tag (BTA-H5258) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession AAP44003.1).This protein carries a mouse IgG 1 * 100 µG
Catalog # ACRBBTA-H5258100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human BTLA (31-150) Protein, Mouse IgG2a Fc Tag,Human BTLA (31-150), Mouse IgG2a Fc Tag (BTA-H5258) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession AAP44003.1).This protein carries a mouse IgG 1 * 100 µG
Catalog # ACRBBTA-H5258100UG
Supplier:  ACRO Biosystems
Frequently Bought Together