Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human S100A8 Protein, Tag Free,Human S100A8, Tag Free (S18-H5112) is expressed from E.coli cells. It contains AA Met 1 - Glu 93 (Accession AAH05928).This protein carries no tag. The protein has a calculated MW of 10.8 kDa. The prot 1 * 100 µG
Catalog # ACRBS18-H5112100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human S100A8 Protein, Tag Free,Human S100A8, Tag Free (S18-H5112) is expressed from E.coli cells. It contains AA Met 1 - Glu 93 (Accession AAH05928).This protein carries no tag. The protein has a calculated MW of 10.8 kDa. The prot 1 * 100 µG
Catalog # ACRBS18-H5112100UG
Supplier:  ACRO Biosystems
Frequently Bought Together