Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human S100A6 Protein, Tag Free,Human S100A6, Tag Free (S16-H5111) is expressed from E.coli cells. It contains AA Met 1 - Gly 90 (Accession AAH01431).This protein carries no tag. The protein has a calculated MW of 10.2 kDa. The prot 1 * 1 mg
Catalog # ACRBS16-H5111-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human S100A6 Protein, Tag Free,Human S100A6, Tag Free (S16-H5111) is expressed from E.coli cells. It contains AA Met 1 - Gly 90 (Accession AAH01431).This protein carries no tag. The protein has a calculated MW of 10.2 kDa. The prot 1 * 1 mg
Catalog # ACRBS16-H5111-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together