Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human MMP-9 Protein, His Tag,Human MMP-9, His Tag (MM9-H5221) is expressed from human 293 cells (HEK293). It cont ains AA Ala 20 - Pro 469 (Accession AAH06093).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 1 mg
Catalog # ACRBMM9-H5221-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human MMP-9 Protein, His Tag,Human MMP-9, His Tag (MM9-H5221) is expressed from human 293 cells (HEK293). It cont ains AA Ala 20 - Pro 469 (Accession AAH06093).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 1 mg
Catalog # ACRBMM9-H5221-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together