Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Cynomolgus Axl Protein, His Tag,Cynomolgus Axl, His Tag (AXL-C52H3) is expressed from human 293 cells (HEK293). I t contains AA Pro 27 - Pro 293 (Accession XP005589384).This protein carries a polyhistidine tag at the C-terminus. Th 1 * 100 µG
Catalog # ACRBAXL-C52H3100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Cynomolgus Axl Protein, His Tag,Cynomolgus Axl, His Tag (AXL-C52H3) is expressed from human 293 cells (HEK293). I t contains AA Pro 27 - Pro 293 (Accession XP005589384).This protein carries a polyhistidine tag at the C-terminus. Th 1 * 100 µG
Catalog # ACRBAXL-C52H3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together