Order Entry
Netherlands
ContactUsLinkComponent
Human PLGF, His Tag (PGF-H5229) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 170 (Accession NP002623.2). 1 * 500 µG
Catalog # ACRBPGF-H5229500UG
Supplier:  ACRO Biosystems
Human PLGF, His Tag (PGF-H5229) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 170 (Accession NP002623.2). 1 * 500 µG
Catalog # ACRBPGF-H5229500UG
Supplier:  ACRO Biosystems
Frequently Bought Together