Chloroform 100 µg/mL in Methanol 1 * 1 mL
Catalog # EHERGA09011107ME
Supplier: EHRENSTORFER
Frequently Bought Together
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
NORVALINE 1 * 50 mg
Supplier: Merck
Gold standard solution, for ICP, in 20% HCl, VHG
Supplier: LGC Standards PROMOCHEM
VOL.FLASK 10ML NS10/19 WIDE GLASS STOPPE 1 * 2 items
Supplier: HIRSCHMANN
Calcium Standard for AAS, TraceCERT TM 1 * 250 mL
Supplier: Merck
TIMOLOL MALEATE 1 * 500 mg
Supplier: Merck
L-Citrulline, Reference Standard
Supplier: USP
(±)-Camphene
Supplier: Merck
Supelco® Organic Reference Standard, Protein Standard Set
Supplier: Merck
MULTIPLE STANDARD SOL. LOW VARION ES/1 1 * 1 L
Supplier: WTW ONLINE
LINEZOLID RELATED COMPOUND D 1 * 50 mg
Supplier: Merck
ATORVASTATIN SYSTEM SUITABILITY 1 * 10 mg
Supplier: EDQM
Yttrium ICP Standards, (CRMs), Fluka™
Supplier: Honeywell Chemicals
MULTI ELEMENT STD 75 BLANK OIL S-21:30 1 * 200 g
Supplier: LGC Standards PROMOCHEM
Buffer Solutions
Supplier: Thermo Fisher Scientific