Order Entry
Netherlands
ContactUsLinkComponent
Stericlin sterilisation double wrap, 2 sheets sealed at the edges, both SMMMS 43g/m², one blue, one green, 120x120cm, DIN EN ISO 116 07 1 * 50 items
Catalogus # VEPA3FDOP381218
Stericlin sterilisation double wrap, 2 sheets sealed at the edges, both SMMMS 43g/m², one blue, one green, 120x120cm, DIN EN ISO 116 07 1 * 50 items
Catalogus # VEPA3FDOP381218
Frequently Bought Together