Order Entry
Netherlands
ContactUsLinkComponent
Stericlin® sterilisation reel, flat, paper 70g/m² with see-through film, with indicators for STEAM, EO and FORM, width 50cm x length 200m, DIN EN ISO 11607 1 * 1 Roll
Catalog # VEPA3FKFS230122
Stericlin® sterilisation reel, flat, paper 70g/m² with see-through film, with indicators for STEAM, EO and FORM, width 50cm x length 200m, DIN EN ISO 11607 1 * 1 Roll
Catalog # VEPA3FKFS230122
Frequently Bought Together